CH6458423858 Chem Impex International, Inc.

Tetrahydropyran-3-ylamine

  • MDL number MFCD07371531
  • Cas Number : 120811-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/tetrahydropyran-3-ylamine/11936?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/tetrahydropyran-3-ylamine/11936?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart