CH6945317377 Chem Impex International, Inc.

DL-Phenylalanine tert-butyl ester hydrochloride

  • MDL number MFCD00190980
  • Cas Number : 75898-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Phe-OtBu HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-phenylalanine-i-tert-i-butyl-ester-hydrochloride/5775?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-phenylalanine-i-tert-i-butyl-ester-hydrochloride/5775?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart