CH6945317390 Chem Impex International, Inc.

2-Amino-5-cyanobenzoic acid

  • MDL number MFCD20485816
  • Cas Number : 99767-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-255
Purity 97
Storage temp. Store at 0-8
Physical Form Pale yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-amino-5-cyanobenzoic-acid/20615?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-amino-5-cyanobenzoic-acid/20615?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart