CH6947293668 Chem Impex International, Inc.

Fmoc-2-chloro-D-b-homophenylalanine

  • MDL number MFCD01860899
  • Cas Number : 268734-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-beta-HomoPhe(2-Cl)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-chloro-d-ß-homophenylalanine/6643?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-chloro-d-ß-homophenylalanine/6643?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart