CH6947293695 Chem Impex International, Inc.

3,3',5,5'-Tetramethylbenzidine dihydrochloride dihydrate

  • MDL number MFCD03065942
  • Cas Number : 207738-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to red powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-3-5-5-tetramethylbenzidine-dihydrochloride/12759?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-3-5-5-tetramethylbenzidine-dihydrochloride/12759?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart