CH6947293894 Chem Impex International, Inc.

2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-galactopyranose

  • MDL number MFCD00050926
  • Cas Number : 76375-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230
Synonym D-Galactosamine pentaacetate
Purity 98
Storage temp. Store under -20
Physical Form White crystal or crystal powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Boiling point 530.163 �t 760 mmHg
Product Link http://www.chemimpex.com/product/productinfo/2-acetamido-1-3-4-6-tetra-o-acetyl-2-deoxy-d-galactopyranose/20808?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-acetamido-1-3-4-6-tetra-o-acetyl-2-deoxy-d-galactopyranose/20808?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart