CH6947293925 Chem Impex International, Inc.

Pentadecanedioic acid

  • MDL number MFCD00039534
  • Cas Number : 1460-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-116
Purity 97
Storage temp. Store at RT
Physical Form Grey to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pentadecanedioic-acid/20848?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pentadecanedioic-acid/20848?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart