CH6947293944 Chem Impex International, Inc.

Chloro-dipiperidinocarbenium hexafluorophosphate

  • MDL number MFCD00191817
  • Cas Number : 161308-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-125
Synonym PipClU
Purity 99
Storage temp. Store at 2-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/chloro-dipiperidinocarbenium-hexafluorophosphate/21005?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/chloro-dipiperidinocarbenium-hexafluorophosphate/21005?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart