CH6947293946 Chem Impex International, Inc.

O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate

  • MDL number MFCD00077730
  • Cas Number : 125700-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-138
Synonym N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl) uroniumtetrafluoroborate
Purity 99
Storage temp. Store at 2-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/-i-o-i-3-4-dihydro-4-oxo-1-2-3-benzotriazin-3-yl-i-n-i-i-n-i-i-n-i-i-n-i-tetramethyluronium-tetrafluoroborate/21007?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/-i-o-i-3-4-dihydro-4-oxo-1-2-3-benzotriazin-3-yl-i-n-i-i-n-i-i-n-i-i-n-i-tetramethyluronium-tetrafluoroborate/21007?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart