CH6947293952 Chem Impex International, Inc.

1-(p-Toluenesulfonyl)-1,2,4-triazole

  • MDL number MFCD00458591
  • Cas Number : 13578-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-110
Synonym 1-Tosyl-1,2,4-triazole
Purity 99
Storage temp. Store at 2-8
Physical Form White crystal
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-p-i-toluenesulfonyl-1-2-4-triazole/21013?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-p-i-toluenesulfonyl-1-2-4-triazole/21013?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart