CH6947293975 Chem Impex International, Inc.

1-(4-Cyanophenyl)guanidine hydrochloride

  • MDL number MFCD16619304
  • Cas Number : 373690-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-240
Synonym N-(4-Cyanophenyl)guanidine hydrochloride
Purity 99
Storage temp. Store at 2-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-4-cyanophenyl-guanidine-hydrochloride/21036?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-4-cyanophenyl-guanidine-hydrochloride/21036?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart