CH6947293982 Chem Impex International, Inc.

N-(4-Cyanophenyl)guanidine

  • MDL number MFCD16251323
  • Cas Number : 5637-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-215
Synonym 1-(4-Cyanophenyl)guanidine
Purity 99
Storage temp. Store at 2-8
Physical Form White crystal
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/-i-n-i-4-cyanophenyl-guanidine/21043?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/-i-n-i-4-cyanophenyl-guanidine/21043?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart