CH6947293984 Chem Impex International, Inc.

Chloroformamidine hydrochloride

  • MDL number MFCD00035527
  • Cas Number : 29671-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-181
Synonym Carbamimidic chloride hydrochloride
Purity 99
Storage temp. Store at 2-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/chloroformamidine-hydrochloride/21045?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/chloroformamidine-hydrochloride/21045?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart