CH6947294013 Chem Impex International, Inc.

1-(4-Pyridyl)pyridinium chloride hydrochloride

  • MDL number MFCD04038041
  • Cas Number : 5421-92-1
  • Molecular Weight : 247.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-160 �dec.)
Purity 99
Storage temp. Store at 2-8
Physical Form Yellow brown powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/1-4-pyridyl-pyridinium-chloride-hydrochloride-hydrate/21074?cid=1

Safety Information

hcodes 8
MSDS http://www.chemimpex.com/product/productinfo/1-4-pyridyl-pyridinium-chloride-hydrochloride-hydrate/21074?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart