CH6947294047 Chem Impex International, Inc.

N,N-Dibenzylglycine ethyl ester

  • MDL number MFCD00796343
  • Cas Number : 77385-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-58
Synonym Ethyl 2-(dibenzylamino)acetate
Purity 0.98
Physical Form White to almost white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-n-dibenzylglycine-ethyl-ester/21108?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-n-dibenzylglycine-ethyl-ester/21108?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart