CH6947294077 Chem Impex International, Inc.

N-Succinimidyl acetate

  • MDL number MFCD00037916
  • Cas Number : 14464-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-140
Synonym Acetic acid N-succinimidyl ester
Purity 99
Storage temp. Store at = - 20 �Protect from Light.
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/-i-n-i-succinimidyl-acetate/21138?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/-i-n-i-succinimidyl-acetate/21138?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart