CH6H11E4EF57 Chem Impex International, Inc.

Acetyl-L-tyrosine methyl ester

  • MDL number MFCD00080844
  • Cas Number : 2440-79-1
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at RT
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-tyrosine-methyl-ester-hydrate/1790?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-tyrosine-methyl-ester-hydrate/1790?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart