CH6H11E4EF58 Chem Impex International, Inc.

O-Benzyl-L-threonine

  • MDL number MFCD00037768
  • Cas Number : 4378-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Thr(Bzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-l-threonine-hydrochloride/2120?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-l-threonine-hydrochloride/2120?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart