CH6H11E4EF9F Chem Impex International, Inc.

D-Glutamic acid gamma-tert-butyl ester alpha-amide hydrochloride

  • MDL number MFCD30491706
  • Molecular Weight : 238.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200 - 206
Purity 99.5
Storage temp. Store at 0 - 8
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/d-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-amide-hydrochloride/20740?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-amide-hydrochloride/20740?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart