CH6H11E4F0EB Chem Impex International, Inc.

Fmoc-cis-4-methylthio-Pro-OH

  • MDL number MFCD30749170
  • Cas Number : 1887209-51-9
  • Molecular Weight : 383.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-cis-i-4-methylthio-pro-oh/20950?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-cis-i-4-methylthio-pro-oh/20950?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart