CH6H11E4F161 Chem Impex International, Inc.

Nalpha-Fmoc-Ndelta-trityl-L-Ornithine

  • MDL number MFCD30478737
  • Cas Number : 1998701-26-0
  • Molecular Weight : 596.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-142
Purity 99.9
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/n-i-sup-alpha-i-sup-fmoc-i-n-i-i-sup-delta-i-sup-trityl-l-ornithine/22138?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-i-sup-alpha-i-sup-fmoc-i-n-i-i-sup-delta-i-sup-trityl-l-ornithine/22138?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart