CH6H11E4F1BF Chem Impex International, Inc.

Pamoic acid disodium salt

  • MDL number MFCD00036183
  • Cas Number : 6640-22-8
  • Molecular Weight : 432.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at RT
Physical Form Pale yellow to yellow powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/pamoic-acid-disodium-salt/21448?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pamoic-acid-disodium-salt/21448?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart