CH6H11E4F279 Chem Impex International, Inc.

Sophoricoside

  • MDL number MFCD01075138
  • Cas Number : 152-95-4
  • Molecular Weight : 432.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 2 - 8
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/sophoricoside/21990?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/sophoricoside/21990?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart