CH6H11E4F38B Chem Impex International, Inc.

H-Gly-Cys-OH

  • MDL number MFCD00672396
  • Cas Number : 57281-78-4
  • Molecular Weight : 178.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 2-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/h-gly-cys-oh/22216?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-gly-cys-oh/22216?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart