CH6H8C2C43E3 Chem Impex International, Inc.

Myristoyl coenzyme A monohydrate

  • MDL number MFCD00150831
  • Cas Number : 3130-72-1
  • Molecular Weight : 995.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Storage temp. Store at = -15 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/myristoyl-coenzyme-a/1209?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/myristoyl-coenzyme-a/1209?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart