CH6H8C2C456D Chem Impex International, Inc.

Fmoc-L-Lys(Boc-AEEA)-OH

  • MDL number MFCD27952843
  • Cas Number : 1662688-16-5
  • Molecular Weight : 613.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75 - 78
Purity 99
Storage temp. Store at 2-8
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-lys-boc-aeea-oh/23135?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-lys-boc-aeea-oh/23135?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart