CH6H8C2C4572 Chem Impex International, Inc.

N-Fmoc-7-methyl-L-tryptophan

  • MDL number MFCD31560627
  • Cas Number : 1808268-53-2
  • Molecular Weight : 440.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99.6
Storage temp. Store at 2 - 8
Physical Form White to off-white powder/solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/-i-n-i-fmoc-7-methyl-l-tryptophan/23219?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/-i-n-i-fmoc-7-methyl-l-tryptophan/23219?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart