CH6H8C2C4575 Chem Impex International, Inc.

Fmoc-2-(2-[2-(2-aminoethoxy)ethoxy]ethoxy)ethanol

  • MDL number MFCD31657165
  • Cas Number : 868594-41-6
  • Molecular Weight : 415.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 2-8
Physical Form Light yellow oil
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2-2-2-2-aminoethoxy-ethoxy-ethoxy-ethanol/23150?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2-2-2-2-aminoethoxy-ethoxy-ethoxy-ethanol/23150?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart