CH6H8C2C4737 Chem Impex International, Inc.

3beta-[N-(N',N'-Dimethylaminoethane)-carbamoyl]cholesterol

  • MDL number MFCD30536817
  • Cas Number : 137056-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-beta-i-n-i-i-n-i-i-n-i-dimethylaminoethane-carbamoyl-cholesterol/5257?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-beta-i-n-i-i-n-i-i-n-i-dimethylaminoethane-carbamoyl-cholesterol/5257?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart