CH6H8C2C4D00 Chem Impex International, Inc.

(R)-4-(Fmoc-amino)-3-phenylbutyric acid

  • MDL number MFCD21363162
  • Cas Number : 1858223-89-8
  • Molecular Weight : 401.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176 - 180
Purity 99
Storage temp. Store at 2-8
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/-i-r-i-4-fmoc-amino-3-phenylbutyric-acid/23132?cid=1

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/-i-r-i-4-fmoc-amino-3-phenylbutyric-acid/23132?cid=1
Availability details for
Quantity
  • Availability
  • Add to Cart