CH6H8C2C4D08 Chem Impex International, Inc.

Ala-Ala-OH HCl

  • MDL number MFCD28515397
  • Cas Number : 1948-31-8
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-ala-oh-font-size-2-face-symbol-middot-font-hcl/2233?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-ala-oh-font-size-2-face-symbol-middot-font-hcl/2233?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart