CH6H8C2C4D09 Chem Impex International, Inc.

Nalpha-Acetyl-L-arginine amide acetate salt

  • MDL number MFCD30187619
  • Cas Number : 88530-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to pale yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-acetyl-l-arginine-amide/1758?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-acetyl-l-arginine-amide/1758?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart