CH6H8C2C4D36 Chem Impex International, Inc.

Nalpha-Nim-Bis-Z-L-histidine�anol

  • MDL number MFCD00153335
  • Molecular Weight : 469.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-n-i-sup-im-sup-i-i-bis-i-i-z-l-histidine/1407?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-n-i-sup-im-sup-i-i-bis-i-i-z-l-histidine/1407?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart