CH6H8C2C4D3C Chem Impex International, Inc.

6-Chloro-3-indolyl-beta-D-galactopyranoside

  • MDL number MFCD01310928
  • Cas Number : 138182-21-5
  • Molecular Weight : 329.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white cream powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-chloro-3-indolyl-beta-d-galactopyranoside/12586?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-chloro-3-indolyl-beta-d-galactopyranoside/12586?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart