CH6H8C2C4D6C Chem Impex International, Inc.

5'-O-(4,4'-dimethoxytrityl)-thymidine-3'-O-succinic acid triethylammonium salt

  • MDL number MFCD12912408

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-thymidine-3-i-o-i-succinic-acid/652?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-thymidine-3-i-o-i-succinic-acid/652?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart