CH6H8C2C4DC1 Chem Impex International, Inc.

Fmoc-S-[(R)-2,3-bis(palmitoyloxy)propyl]-L-cysteine

  • MDL number MFCD30478735
  • Cas Number : 139573-77-6
  • Molecular Weight : 894.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 90
Purity 99
Storage temp. Store at = -20
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/fmoc-i-s-i-i-r-i-2-3-bis-palmitoyloxy-propyl-l-cysteine

Safety Information

MSDS http://www.chemimpex.com/fmoc-i-s-i-i-r-i-2-3-bis-palmitoyloxy-propyl-l-cysteine
Availability details for
Quantity
  • Availability
  • Add to Cart