CH6H8C2C4DC2 Chem Impex International, Inc.

5(6)-Carboxyfluorescein

  • MDL number MFCD31693849
  • Cas Number : 72088-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0 - 8 °C
Physical Form Burnt orange or orange to yellow brown so
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-6-carboxyfluorescein/383?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-6-carboxyfluorescein/383?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart