COM448611077 Combi-Blocks, Inc.

(Tetrahydropyran-4-ylidene)acetonitrile

  • MDL number MFCD09800849
  • Cas Number : 204651-40-1
  • Molecular Weight : 123.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1621
Availability details for
Quantity
  • Availability
  • Add to Cart