COM448611120 Combi-Blocks, Inc.

4,5,6,7-Tetrahydro-1H-indazole HCl

  • MDL number MFCD13195636
  • Cas Number : 18161-11-0
  • Molecular Weight : 158.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1740
Availability details for
Quantity
  • Availability
  • Add to Cart