COM448611183 Combi-Blocks, Inc.

1-benzylpiperidin-3-one

  • MDL number MFCD00023277
  • Cas Number : 40114-49-6
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1831

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1831
Availability details for
Quantity
  • Availability
  • Add to Cart