COM448611188 Combi-Blocks, Inc.

piperidin-3-ylmethanol, HCl

  • MDL number MFCD08752661
  • Cas Number : 400771-49-5
  • Molecular Weight : 151.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1837

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AM-1837
Availability details for
Quantity
  • Availability
  • Add to Cart