COM448611285 Combi-Blocks, Inc.

(3R)-N-methylpyrrolidin-3-amine

  • MDL number MFCD00191461
  • Cas Number : 139015-33-1
  • Molecular Weight : 100.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2032

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2032
Availability details for
Quantity
  • Availability
  • Add to Cart