COM448611286 Combi-Blocks, Inc.

1-benzyl-N,N-dimethylpyrrolidin-3-amine

  • MDL number MFCD00191304
  • Cas Number : 69478-77-9
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2036

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2036
Availability details for
Quantity
  • Availability
  • Add to Cart