COM448611305 Combi-Blocks, Inc.

Methyl 4-(4-bromophenyl)piperazine-1-carboxylate

  • MDL number MFCD16036098
  • Cas Number : 841295-69-0
  • Molecular Weight : 299.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2073
Availability details for
Quantity
  • Availability
  • Add to Cart