COM448611306 Combi-Blocks, Inc.

4-(4-Isopropoxycarbonyl)piperazino-1-bromobenzene

  • MDL number MFCD16295048
  • Cas Number : 1226808-58-7
  • Molecular Weight : 327.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2074
Availability details for
Quantity
  • Availability
  • Add to Cart