COM448611307 Combi-Blocks, Inc.

1-(4-Bromophenyl)-4-(methylsulfonyl)piperazine

  • MDL number MFCD14584697
  • Cas Number : 357647-98-4
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2075
Availability details for
Quantity
  • Availability
  • Add to Cart