COM448611310 Combi-Blocks, Inc.

4-(4-Bromophenyl)-N,N-dimethylpiperazine-1-sulfonamide

  • MDL number MFCD16295050
  • Cas Number : 1226808-59-8
  • Molecular Weight : 348.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2078
Availability details for
Quantity
  • Availability
  • Add to Cart