COM448611311 Combi-Blocks, Inc.

Cbz-L-Tyrosine benzyl ester triflate

  • MDL number MFCD16295051
  • Cas Number : 183070-41-9
  • Molecular Weight : 537.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2079
Availability details for
Quantity
  • Availability
  • Add to Cart