COM448611337 Combi-Blocks, Inc.

N-[1-(4-Bromophenyl)cyclopropyl]acetamide

  • MDL number MFCD19981500
  • Cas Number : 1330750-29-2
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2110
Availability details for
Quantity
  • Availability
  • Add to Cart