COM448611352 Combi-Blocks, Inc.

1-benzyl 3-ethyl pyrrolidine-1,3-dicarboxylate

  • MDL number MFCD09953274
  • Cas Number : 848413-99-0
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2136
Availability details for
Quantity
  • Availability
  • Add to Cart